$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC L-Ornithine L-Aspartate Salt
TRC L-Ornithine L-Aspartate Salt
:
LGC Standards
New Product
TRC L-Ornithine L-Aspartate Salt
:
LGC Standards
New Product
CAS No.:
3230-94-2