$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Perazine-d8 Dihydrochloride Salt
TRC Perazine-d8 Dihydrochloride Salt
77991-428
:
LGC Standards
New Product
:
TRC Perazine-d8 Dihydrochloride Salt
77991-428
:
LGC Standards
:
TRC-P285802-1MG
New Product
:
CAS No.:
1246815-57-5
Description:
Perazine-d8 Dihydrochloride Salt
Size:
1 mg