Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 5-(N-Ethyl-N-isopropyl) Amiloride
TRC 5-(N-Ethyl-N-isopropyl) Amiloride
TRC 5-(N-Ethyl-N-isopropyl) Amiloride
77984-397
:
LGC Standards
New Product
:
TRC 5-(N-Ethyl-N-isopropyl) Amiloride
77984-397
:
LGC Standards
:
TRC-E921500-25MG
New Product
:
CAS No.:
1154-25-2
Description:
5-(N-Ethyl-N-isopropyl) Amiloride
Size:
25 mg