ContactUsLinkComponent
Chlorine Dioxide Neutralizer Solution Pack
Chlorine Dioxide Neutralizer Solution Pack
Catalog # 77674-544
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Chlorine Dioxide Neutralizer Solution Pack
Catalog # 77674-544
Supplier:  CHEMetrics
Supplier Number:  A-2700
New Product
CAS Number:  
Specifications
  • Chemical method:
    DPD
Specifications
About this item
The Chromate Acidifier Solution A-2700 accessory pack supplies enough solution to perform at least 200 tests. Qty. per Pack: six 10 ml bottles.