$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Isomannide Dinitrate
TRC Isomannide Dinitrate
Catalog # 77987-371
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Isomannide Dinitrate
Catalog # 77987-371
Supplier:
LGC Standards
Supplier Number:
TRC-I821335-100MG
New Product
CAS Number:
Specifications
CAS No.:
551-43-9
Description:
Isomannide Dinitrate
Size:
100 mg
Specifications
About this item
Product Details
Product Details