Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC L-Pyroglutamic Acid
TRC L-Pyroglutamic Acid
TRC L-Pyroglutamic Acid
77992-450
:
LGC Standards
New Product
:
TRC L-Pyroglutamic Acid
77992-450
:
LGC Standards
:
TRC-P997480-10G
New Product
:
CAS No.:
98-79-3
Description:
L-Pyroglutamic Acid
Size:
10 g