Order Entry
Puerto Rico
ContactUsLinkComponent
CHEMets® Hydrazine Visual Test Kit 0 to 0.5 ppm
CHEMets® Hydrazine Visual Test Kit 0 to 0.5 ppm
Catalog # 77674-780
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
CHEMets® Hydrazine Visual Test Kit 0 to 0.5 ppm
Catalog # 77674-780
Supplier:  CHEMetrics
Supplier Number:  K-5005
New Product
CAS Number:  

Specifications

  • Chemical method:
    PDMAB
  • Application:
    Boiler feed water;Process water

Specifications

About this item

  • Range: 0 to 0.5 ppm MDL: 0.005 ppm Method: PDMAB
Delivery: K-5005 Kit comes in a plastic case and contains everything needed to perform 30 tests:refill, Comparator, 25 ml sample cup, and instructions.