Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC p-Chlonixin
TRC p-Chlonixin
77980-512
:
LGC Standards
New Product
:
TRC p-Chlonixin
77980-512
:
LGC Standards
:
TRC-C587145-100MG
New Product
:
CAS No.:
36752-40-6
Description:
p-Chlonixin
Size:
100 mg