$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC 1,2-Diethoxyethane
TRC 1,2-Diethoxyethane
:
LGC Standards
New Product
TRC 1,2-Diethoxyethane
:
LGC Standards
New Product
CAS No.:
629-14-1