TRC 1,3-Dimethylpentylamine Hydrochloride Salt
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:13803-74-2
Specifications
Frequently Bought Together
PEEK Tubing, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
Manual Crimpers and Decappers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® Flat-Bottom Fittings, For Tubing Ø > ¹/₁₆" (1.6 mm) and ≤ ¹/₈" (3.2 mm), IDEX Health & Science
Supplier: Upchurch Scientific
Upchurch Scientific® Stainless Steel Frits, IDEX Health & Science
Supplier: Upchurch Scientific
Vaplock™ Chemical Exhaust Filters and Color Indicators, Cole-Parmer
Supplier: Antyila Scientific
Accessories for Gas Clean Purification System, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Captiva Filter Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Source Lamps for UV-Vis & UV-Vis-NIR, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Tubing for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
Supplier: Thermo Scientific
Captiva Syringe Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Solvent Bottles and Waste Cans for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
ZORBAX, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Merlin Microseal for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Detectors for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
J.T.Baker®, Phase Separation Paper
Supplier: AVANTOR PERFORMANCE MATERIALS US
Tubing Kits for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD)