Order Entry
Puerto Rico
ContactUsLinkComponent
Paragon Scientific Base Number, 6.0 mg KOH/g Certified Value
Paragon Scientific Base Number, 6.0 mg KOH/g Certified Value
  77976-027
New Product
 :  
undefined
Paragon Scientific Base Number, 6.0 mg KOH/g Certified Value
  77976-027
 :  ALK-TBN6/3
New Product
 :  

 

  • Pack type:
    Glass bottle
  • Description:
    Base number, 6.0 mg KOH/g certified value
  • Size:
    3×50 g
  • Storage Temperature:
    RT

 

 

  • Shipping conditions: Room temperature