$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC rac-cis-2,3-Dichloro Sertraline Hydrochloride
TRC rac-cis-2,3-Dichloro Sertraline Hydrochloride
77982-402
:
LGC Standards
New Product
:
TRC rac-cis-2,3-Dichloro Sertraline Hydrochloride
77982-402
:
LGC Standards
:
TRC-D435880-10MG
New Product
:
CAS No.:
1198084-29-5
Description:
rac-cis-2,3-Dichloro Sertraline Hydrochloride
Size:
10 mg