Paragon Scientific Bromine Index - 100 mg/100 g
Catalog # 77975-510
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:Bromine index - 100 mg/100 g
- Size:200 ml
- Storage Temperature:RT
Specifications
About this item
- Shipping conditions: Room temperature