Order Entry
Puerto Rico
ContactUsLinkComponent
Chlorine Comparator Round 0 to 1 ppm
Chlorine Comparator Round 0 to 1 ppm
Catalog # 77674-596
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Chlorine Comparator Round 0 to 1 ppm
Catalog # 77674-596
Supplier:  CHEMetrics
Supplier Number:  C-2504
New Product
CAS Number:  

Specifications

  • Chemical method:
    DPD (free and total)

Specifications

About this item

C-2504 Low Range Chlorine Comparator is a component of K-2504 Chlorine (free and total) CHEMets Visual Test Kit and is designed to be compared against the color formed in R-2500 Chlorine CHEMets Refill The comparator's range is applicable for many drinking water and disinfection applications. Color standards are pink in color and cover a range of 0 to 1.0 ppm chlorine (Cl₂).