Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC N-Demethyl Vandetanib
TRC N-Demethyl Vandetanib
TRC N-Demethyl Vandetanib
77981-440
:
LGC Standards
New Product
:
TRC N-Demethyl Vandetanib
77981-440
:
LGC Standards
:
TRC-D231355-50MG
New Product
:
CAS No.:
338992-12-4
Description:
N-Demethyl Vandetanib
Size:
50 mg