Paragon Scientific IP 336, Sulfur set C in heavy mineral oil: S @ 0.5, 1, 1.5, 2%
Catalog # 77975-803
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Heavy Mineral Oil
- CAS No.:7704-34-9
- Description:IP 336, Sulfur set C in heavy mineral oil: S @ 0.5, 1, 1.5, 2%
- Size:4×100 ml
- Certification:ISO 17034
- Storage Conditions:RT
- Shipping Conditions:RT
- Molecular Weight:32.065