ContactUsLinkComponent
CHEMets® Iron In Brine Visual Test Kit
CHEMets® Iron In Brine Visual Test Kit
Catalog # 77674-800
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
CHEMets® Iron In Brine Visual Test Kit
Catalog # 77674-800
Supplier:  CHEMetrics
Supplier Number:  K-6002
New Product
CAS Number:  
Specifications
  • Chemical method:
    Ferric thiocyanate (Iron in brine)
Specifications
About this item
  • Range: 0 to 100 and 100 to 1000 mg/L MDL: 5 mg/L Method: Ferric Thiocyanate (Iron in brine)
Delivery: K-6002 Kit comes in a plastic case and contains everything needed to perform 30 tests (except distilled water):refill, low and high range comparators, acidifier solution, activator solution, 50 ml sample cup with cap, 1.0 ml syringes, and instructions.