TRC Ethyl Loflazepate (1.0 mg/ml in Acetonitrile)
Catalog # 77987-748
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Acetonitrile
- CAS No.:29177-84-2
- Description:Ethyl Loflazepate (1.0 mg/ml in Acetonitrile)
- Size:1×1ml