$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC N-Oleoylethanolamide-d4
TRC N-Oleoylethanolamide-d4
77990-828
:
LGC Standards
New Product
:
TRC N-Oleoylethanolamide-d4
77990-828
:
LGC Standards
:
TRC-O527282-5MG
New Product
:
CAS No.:
946524-36-3
Description:
N-Oleoylethanolamide-d4
Size:
5 mg