Paragon Scientific 10W30 Viscosity Check Oil (Kinematic Viscosity, CCS Dynamic Viscosity and Density)
Supplier: LGC Standards
New Product
Certificates
About this item
- Standard type: Single component
Specifications
- Shipping Conditions:RT
- Storage Temperature:RT