Order Entry
Puerto Rico
ContactUsLinkComponent
Paragon Scientific Color Standard USP (631) Ferric Chloride CS
Paragon Scientific Color Standard USP (631) Ferric Chloride CS
Catalog # 77976-032
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Paragon Scientific Color Standard USP (631) Ferric Chloride CS
Catalog # 77976-032
Supplier:  LGC Standards
Supplier Number:  ALK-USPPRYE
New Product
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Description:
    Colour standard USP (631) Ferric Chloride CS
  • Size:
    100 ml
  • Storage temperature:
    RT

Specifications

About this item

  • Matrix: Aq. HCl