$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC DL-Serine-2,3,3-d3
TRC DL-Serine-2,3,3-d3
Catalog # 77993-105
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC DL-Serine-2,3,3-d3
Catalog # 77993-105
Supplier:
LGC Standards
Supplier Number:
TRC-S270982-50MG
New Product
CAS Number:
Specifications
CAS No.:
70094-78-9
Description:
DL-Serine-2,3,3-d3
Size:
50 mg
Specifications
About this item
Product Details
Product Details