TRC Azithromycin N-Oxide
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:90503-06-3
Specifications
Frequently Bought Together
Manual Crimpers and Decappers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® PEEK™ Tubing, IDEX Health & Science
Supplier: Upchurch Scientific
Upchurch Scientific® Luer Adapters, IDEX Health & Science
Supplier: Upchurch Scientific
PEEK Tubing, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
Upchurch Scientific® Flat-Bottom Fittings, For Tubing Ø > ¹/₁₆" (1.6 mm) and ≤ ¹/₈" (3.2 mm), IDEX Health & Science
Supplier: Upchurch Scientific
Vaplock™ Chemical Exhaust Filters and Color Indicators, Cole-Parmer
Supplier: Antyila Scientific
CrossLab Instrument Supplies for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Solvent Bottles and Waste Cans for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
Supplier: Thermo Scientific
Stainless Steel Frits, Upchurch Scientific®
Supplier: Upchurch Scientific
Agilent Autosampler Supplies, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Captiva Syringe Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Sample Trays for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Baskets, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
InfinityLab Well Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
ZORBAX, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)