Orders LinkContactUsLinkComponent
DEHA Activator Solution Pack
DEHA Activator Solution Pack
PDTS
Catalog # 77674-550
Supplier:   CHEMetrics
Supplier #:  A-3900
DEHA Activator Solution Pack
Catalog # 77674-550
Supplier:  CHEMetrics
CAS Number:  
DEHA Activator Solution Pack
DEHA Activator Solution Pack
PDTS
CAS Number:  
Price
$16.97
Log in for availability
Pack of 6 (10ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The accessory pack A-3900 supplies enough DEHA activator solution to perform at least 200 tests. Qty. per Pack:Six 10 ml Bottles.
Specifications
Specifications
  • Chemical method:
    PDTS