Order Entry
Puerto Rico
ContactUsLinkComponent
Paragon Scientific Cone and Plate (Nominal: 50 mPas @ 25 °C), Viscosity
Paragon Scientific Cone and Plate (Nominal: 50 mPas @ 25 °C), Viscosity
  77975-467
New Product
 :  
undefined
Paragon Scientific Cone and Plate (Nominal: 50 mPas @ 25 °C), Viscosity
  77975-467
 :  ALK-2162/1
New Product
 :  

 

  • Description:
    Cone and plate (nominal: 50 mPas @ 25 °C), viscosity
  • Size:
    500 ml
  • Storage Temperature:
    RT
  • Certification:
    ISO 17034

 

 

  • Standard type: Single component