$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC 4-Desmethoxy-4-chloro Omeprazole
TRC 4-Desmethoxy-4-chloro Omeprazole
77981-807
:
LGC Standards
New Product
:
TRC 4-Desmethoxy-4-chloro Omeprazole
77981-807
:
LGC Standards
:
TRC-D290390-10MG
New Product
:
CAS No.:
863029-89-4
Description:
4-Desmethoxy-4-chloro Omeprazole
Size:
10 mg