$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Bisphenol AP
TRC Bisphenol AP
77978-753
:
LGC Standards
New Product
:
TRC Bisphenol AP
77978-753
:
LGC Standards
:
TRC-B519485-5G
New Product
:
CAS No.:
1571-75-1
Description:
Bisphenol AP
Size:
5 g