Dr. Ehrenstorfer Microplastics Polyethylene PE nominal 50 μm) (Dv50 = 120 μm)
Catalog # 77976-075
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:9002-88-4
- Description:Microplastics polyethylene PE nominal 50 μm (Dv50 = 120 μm; Dn50 = 3 μm)
- Size:50 mg
- Shipping Conditions:20 °C
- Formula:C₂H₄
- Formula Weight:28,053 g/mol
- Storage Temperature:20 °C
- Certification:ISO/IEC 17025