$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Beta-Sitosterol (Synthetic)
TRC Beta-Sitosterol (Synthetic)
:
LGC Standards
New Product
TRC Beta-Sitosterol (Synthetic)
:
LGC Standards
New Product
CAS No.:
83-46-5