$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC sn-Glycero-3-phosphocholine-d9
TRC sn-Glycero-3-phosphocholine-d9
TRC sn-Glycero-3-phosphocholine-d9
Catalog # 77985-600
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC sn-Glycero-3-phosphocholine-d9
Catalog # 77985-600
Supplier:
LGC Standards
Supplier Number:
TRC-G598702-10MG
New Product
CAS Number:
Specifications
CAS No.:
2260669-07-4
Description:
sn-Glycero-3-phosphocholine-d9
Size:
10 mg
Specifications
About this item
Product Details
Product Details