Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 3-Hydroxy-4-morpholino-1,2,5-thiazole
TRC 3-Hydroxy-4-morpholino-1,2,5-thiazole
TRC 3-Hydroxy-4-morpholino-1,2,5-thiazole
77986-663
:
LGC Standards
New Product
:
TRC 3-Hydroxy-4-morpholino-1,2,5-thiazole
77986-663
:
LGC Standards
:
TRC-H948475-250MG
New Product
:
CAS No.:
30165-97-0
Description:
3-Hydroxy-4-morpholino-1,2,5-thiazole
Size:
250 mg