Order Entry
Puerto Rico
ContactUsLinkComponent
Nitrite Instrumental Test 0 to 0.750 ppm
Nitrite Instrumental Test 0 to 0.750 ppm
Catalog # 77674-836
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Nitrite Instrumental Test 0 to 0.750 ppm
Catalog # 77674-836
Supplier:  CHEMetrics
Supplier Number:  K-7013
New Product
CAS Number:  

Specifications

  • Chemical method:
    Azo dye formation
  • Application:
    Industrial waste water influent and effluent;Industrial process water;Boiler water;Cooling water;Surface and ground water

Specifications

About this item

  • Range: 0 to 0.750 ppm as N Method: Azo Dye Formation (NED)
Delivery: K-7013 Nitrite Vacu-vials test kit is used to measure the concentration of nitrite in water with the azo dye formation method. It comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, acidifier solution, 25 ml sample cup, ampoule blank, and instructions. This kit requires the use of a CHEMetrics direct-readout photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.