Order Entry
Puerto Rico
ContactUsLinkComponent
TRC 2-(4-Aminophenyl)-1,2-dihydro-1-oxo-7-(2-pyridinylmethoxy)-4-(3,4,5-trimethoxyphenyl)-3-isoquinolinecarboxylic Acid Methyl Ester Dihydrochloride (>90%)
TRC 2-(4-Aminophenyl)-1,2-dihydro-1-oxo-7-(2-pyridinylmethoxy)-4-(3,4,5-trimethoxyphenyl)-3-isoquinolinecarboxylic Acid Methyl Ester Dihydrochloride (>90%)
Catalog # 77977-330
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
TRC 2-(4-Aminophenyl)-1,2-dihydro-1-oxo-7-(2-pyridinylmethoxy)-4-(3,4,5-trimethoxyphenyl)-3-isoquinolinecarboxylic Acid Methyl Ester Dihydrochloride (>90%)
Catalog # 77977-330
Supplier:  LGC Standards
Supplier Number:  TRC-A622635-10MG
New Product
CAS Number:  

Specifications

  • CAS No.:
    212498-37-8
  • Description:
    2-(4-Aminophenyl)-1,2-dihydro-1-oxo-7-(2-pyridinylmethoxy)-4-(3,4,5-trimethoxyphenyl)-3-isoquinolinecarboxylic Acid Methyl Ester Dihydrochloride (>90%)
  • Size:
    10 mg

Specifications

About this item