Order Entry
Puerto Rico
ContactUsLinkComponent
Paragon Scientific Viscosity Set, Cold Crank Simulator Type: CL10, CL12, CL14, CL16, CL19, CL22, CL25 (2) , CL28, CL32, CL38, CL48, CL60, CL68
Paragon Scientific Viscosity Set, Cold Crank Simulator Type: CL10, CL12, CL14, CL16, CL19, CL22, CL25 (2) , CL28, CL32, CL38, CL48, CL60, CL68
Catalog # 77976-033
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Paragon Scientific Viscosity Set, Cold Crank Simulator Type: CL10, CL12, CL14, CL16, CL19, CL22, CL25 (2) , CL28, CL32, CL38, CL48, CL60, CL68
Catalog # 77976-033
Supplier:  LGC Standards
Supplier Number:  ALK-VIS-CCS-14
New Product
CAS Number:  

Specifications

  • Description:
    Viscosity set, cold crank simulator type: CL10, CL12, CL14, CL16, CL19, CL22, CL25 (2) , CL28, CL32, CL38, CL48, CL60, CL68
  • Size:
    14×500 ml
  • Shipping Conditions:
    RT
  • Storage Temperature:
    RT
  • Certification:
    ISO 17034

Specifications

About this item

  • Standard type: Single component