ContactUsLinkComponent
CHEMets® Iron Total Soluble HR Visual Test Kit 10 to 100 ppm
CHEMets® Iron Total Soluble HR Visual Test Kit 10 to 100 ppm
  77674-806
New Product
 :  
undefined
CHEMets® Iron Total Soluble HR Visual Test Kit 10 to 100 ppm
  77674-806
 :  K-6020D
New Product
 :  
  • Chemical method:
    Phenanthroline (total and soluble)
  • Application:
    Industrial waste water influent and effluent;Industrial process water;Surface and ground water;Sea water;Drinking water
The K-6020D Iron (total and soluble) HR CHEMets® test kit (10 to 100 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): HR CHEMets® refill, flat comparator, 25 ml sample cup, 3.0 ml syringe, iron activator solution, and instructions. Note: Kit contains only enough activator solution to perform approximately 7 total iron tests.
  • Method: Phenanthroline (total and soluble) Range: 10 to 100 ppm MDL: 10 ppm
 : The K-6020D Iron (total and soluble) HR CHEMets® test kit (10 to 100 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): HR CHEMets®refill, flat comparator, 25 ml sample cup, 3.0 ml syringe, iron activator solution, and instructions. Note: Kit contains only enough activator solution to perform approximately 7 total iron tests.