$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Lipomed (-)-Cotinine.HCl, 1 mg/ml in Methanol (as free base)
Lipomed (-)-Cotinine.HCl, 1 mg/ml in Methanol (as free base)
77976-132
:
LGC Standards
New Product
:
Lipomed (-)-Cotinine.HCl, 1 mg/ml in Methanol (as free base)
77976-132
:
LGC Standards
:
LPM-COT-1403-HC-1LM
New Product
:
Matrix:
Methanol
Description:
(-)-Cotinine.HCl, 1 mg/ml in Methanol (as free base)
Size:
1 ml