Paragon Scientific 147 µS/cm @ 25 °C, Conductivity (NIST Traceable, ACS)
Catalog # 77975-549
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:Conductivity standard CRM
- Size:500 ml
- Reference temperature:25 °C
- Certification:ISO 17034
- Conductivity:147 µS/cm