$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC 3-Bromo-1,2-propanediol
TRC 3-Bromo-1,2-propanediol
77979-008
:
LGC Standards
New Product
:
TRC 3-Bromo-1,2-propanediol
77979-008
:
LGC Standards
:
TRC-B686655-1G
New Product
:
CAS No.:
4704-77-2
Description:
3-Bromo-1,2-propanediol
Size:
1 g