Paragon Scientific IP 496, Sulfur set A in synthetic diesel: S @ blank, 0.01, 0.03, 0.05, 0.07, 0.1%
77975-819
New Product
:
- Matrix:Synthetic Diesel
- CAS No.:7704-34-9
- Description:IP 496, Sulfur set A in synthetic diesel: S @ blank, 0.01, 0.03, 0.05, 0.07, 0.1%
- Size:6×100ml
- Certification:ISO 17034
- Storage Conditions:RT
- Shipping Conditions:RT
- Molecular Weight:32.065