Order Entry
Puerto Rico
ContactUsLinkComponent
 
CHEMets® Phosphate Visual Test Kit 0 to 120 ppm
Catalog # 77674-862
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
CHEMets® Phosphate Visual Test Kit 0 to 120 ppm
Catalog # 77674-862
Supplier:  CHEMetrics
Supplier Number:  K-8515
New Product
CAS Number:  

Specifications

  • Chemical method:
    Vanadomolybdophosphoric Acid
  • Cat. No.:
    77674-862
  • Application:
    Domestic and industrial waste water;Industrial process water;Boiler water;Cooling water;Surface and ground water;Potable water;Sea water;Power generation;Pulp and paper industries;Food and beverages
  • Range:
    10, 20, 30, 40, 60, 80, 100, 120, 150 ppm

Specifications

About this item

  • Range: 0 to 120 ppm MDL: 5 ppm Method: Vanadomolybdophosphoric acid
Delivery: K-8515 Kit comes in a plastic case and contains everything needed to perform 30 tests:refill, comparator, 25 ml sample cup, and instructions.