TRC N-Nitroso-di-n-butylamine-d18 (1.0 mg/ml in Methanol)
Catalog # 77987-780
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- CAS No.:1219798-82-9
- Description:N-Nitroso-di-n-butylamine-d18 (1.0 mg/ml in Methanol)
- Size:1×1ml