Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC o-Aminoazotoluene
TRC o-Aminoazotoluene
TRC o-Aminoazotoluene
Catalog # 77977-045
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC o-Aminoazotoluene
Catalog # 77977-045
Supplier:
LGC Standards
Supplier Number:
TRC-A586000-25G
New Product
CAS Number:
Specifications
CAS No.:
97-56-3
Description:
o-Aminoazotoluene
Size:
25 g
Specifications
About this item
Product Details
Product Details