$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Simazine-d10
TRC Simazine-d10
TRC Simazine-d10
Catalog # 77993-170
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Simazine-d10
Catalog # 77993-170
Supplier:
LGC Standards
Supplier Number:
TRC-S465902-5MG
New Product
CAS Number:
Specifications
CAS No.:
220621-39-6
Description:
Simazine-d10
Size:
5 mg
Specifications
About this item
Product Details
Product Details