Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 5-Methyl Cytosine-13C,15N2 Hydrochloride
TRC 5-Methyl Cytosine-13C,15N2 Hydrochloride
TRC 5-Methyl Cytosine-13C,15N2 Hydrochloride
77988-895
:
LGC Standards
New Product
:
TRC 5-Methyl Cytosine-13C,15N2 Hydrochloride
77988-895
:
LGC Standards
:
TRC-M294702-1MG
New Product
:
CAS No.:
1246815-59-7
Description:
5-Methyl Cytosine-13C,15N2 Hydrochloride
Size:
1 mg