Order Entry
Puerto Rico
ContactUsLinkComponent
 
Paragon Scientific Cone and Plate (Nominal: 350 mPas @ 25 °C), Viscosity
  77975-488
New Product
 :  
undefined
Paragon Scientific Cone and Plate (Nominal: 350 mPas @ 25 °C), Viscosity
  77975-488
 :  ALK-2162/8
New Product
 :  

 

  • Description:
    Cone and plate (nominal: 350 mPas @ 25 °C), viscosity
  • Size:
    500 ml
  • Shipping Conditions:
    RT
  • Storage Temperature:
    RT
  • Certification:
    ISO 17034

 

 

  • Standard type: Single component