ContactUsLinkComponent
CHEMets® Hydrogen Peroxide HR Visual Test Kit 25 to 250 ppm
CHEMets® Hydrogen Peroxide HR Visual Test Kit 25 to 250 ppm
  77674-788
New Product
 :  
undefined
CHEMets® Hydrogen Peroxide HR Visual Test Kit 25 to 250 ppm
  77674-788
 :  K-5520A
New Product
 :  
  • Chemical method:
    Ferric thiocyanate
  • Application:
    Industrial effluent;Waste water;Sea water;Aseptic packaging;Food and beverages
 : The K-5520A hydrogen peroxide HR CHEMets® test kit (25 to 250 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): R to 5510 CHEMets®refill, C to 5510 Comparator, 25 ml sample cup, A to 0027 1.0 ml syringe, and instructions.