TRC 2,2-Dichloro-propanoyl Chloride
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:26073-26-7
Specifications
Frequently Bought Together
PEEK Tubing, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
Upchurch Scientific® PEEK™ Tubing, IDEX Health & Science
Supplier: Upchurch Scientific
Manual Crimpers and Decappers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® Flat-Bottom Fittings, For Tubing Ø > ¹/₁₆" (1.6 mm) and ≤ ¹/₈" (3.2 mm), IDEX Health & Science
Supplier: Upchurch Scientific
Upchurch Scientific® Stainless Steel Frits, IDEX Health & Science
Supplier: Upchurch Scientific
Vaplock™ Chemical Exhaust Filters and Color Indicators, Cole-Parmer
Supplier: Antyila Scientific
Captiva Syringe Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Full Flow Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® Precolumn Filters, IDEX Health & Science
Supplier: Upchurch Scientific
Upchurch Scientific® Flat-Bottom Fittings, For Tubing Ø = ¹/₁₆" (1.6 mm), IDEX Health & Science
Supplier: Upchurch Scientific
CrossLab Solvent Bottles and Waste Cans for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® ETFE Fluoropolymer Tubing, IDEX Health & Science
Supplier: Upchurch Scientific
pH Gradient Buffers
Supplier: Thermo Scientific
Agilent Prep HPLC Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Pump Valves and Supplies for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)