Order Entry
Puerto Rico
ContactUsLinkComponent
CHEMets® Ammonia Visual HR Test Kit (Nessler 25 to 250 ppm)
CHEMets® Ammonia Visual HR Test Kit (Nessler 25 to 250 ppm)
Catalog # 77674-724
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
CHEMets® Ammonia Visual HR Test Kit (Nessler 25 to 250 ppm)
Catalog # 77674-724
Supplier:  CHEMetrics
Supplier Number:  K-1520A
New Product
CAS Number:  

Specifications

  • Chemical method:
    Direct Nesslerization
  • Application:
    Drinking water;Clean surface water;Waste water;Storm water

Specifications

About this item

Delivery: The K-1520A ammonia HR CHEMets® test kit (25 to 250 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): 30 HR CHEMets® ampoules, a color comparator, 1.0 ml syringe, 25 ml sample cup, and instructions.
Caution: Contains mercury. Dispose according to local, state or federal laws.