Nitrate Instrumental Test Zinc Reduction
Catalog # 77674-826
Supplier: CHEMetrics
New Product
CAS Number:
Specifications
- Chemical method:Zinc reduction
- Application:Industrial waste water influent and effluent;Industrial process water;Boiler water;Cooling water;Surface and ground water;Potable water
Specifications
About this item
- Range: 0 to 1.50 ppm as N Method: Zinc reduction
Delivery: Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, thirty zinc foil packs, acidifier solution, reaction tube and cap, 25 ml sample cup, ampoule blank, instructions, and MSDS. This kit requires the use of a CHEMetrics direct-readout photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.