Copper Instrumental Test
Catalog # 77674-766
Supplier: CHEMetrics
New Product
CAS Number:
Specifications
- Chemical method:Bathocuproine
- Description:Vacu-vials kit, copper
- Cat. No.:77674-766
- Application:Drinking water;Ground water;Surface water;Seawater;Waste water
- Range:0 - 12.00 ppm
- No. of tests:30
Specifications
About this item
- Range: V-2000 and V-3000: 0 to 12.00 ppm / Spec: 0 to 7.00 ppm Method: Bathocuproine
Delivery: Kit K-3503 comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, 25 ml sample cup, ampoule blank, and instructions. This kit requires the use of a CHEMetrics Direct to readout photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.